missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-aminopropyl)phenylphosphine, Thermo Scientific Chemicals

Catalog Number p-7048404
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
6775-01-5
C12H21N2P
224.29
MFCD00014834
OEHBMEXOPYWPKQ-UHFFFAOYSA-N
bis 3-aminopropyl phenylphosphine, 3,3'-phenylphosphinediyl bis propan-1-amine, 3-3-aminopropyl phenyl phosphanyl propan-1-amine, bis 3-aminopropyl phenyl phosphane, 1-propanamine,3,3'-phenylphosphinidene bis-9ci, bis 3-aminopropyl phenylphosphine, packaged under argon in resealable chemseal bottles
138827
3-[3-aminopropyl(phenyl)phosphanyl]propan-1-amine
NCCCP(CCCN)C1=CC=CC=C1
This item is not returnable. View return policy
6775-01-5
C12H21N2P
224.29
MFCD00014834
OEHBMEXOPYWPKQ-UHFFFAOYSA-N
bis 3-aminopropyl phenylphosphine, 3,3'-phenylphosphinediyl bis propan-1-amine, 3-3-aminopropyl phenyl phosphanyl propan-1-amine, bis 3-aminopropyl phenyl phosphane, 1-propanamine,3,3'-phenylphosphinidene bis-9ci, bis 3-aminopropyl phenylphosphine, packaged under argon in resealable chemseal bottles
138827
3-[3-aminopropyl(phenyl)phosphanyl]propan-1-amine
NCCCP(CCCN)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.03g/mL
Boiling Point 144°C (1 mmHg)
Refractive Index 1.5728
Quantity 2 g
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 224.29
Physical Form Liquid
Chemical Name or Material Bis(3-aminopropyl)phenylphosphine
compliance-icons
Product Identifier
  • Bis(3-aminopropyl)phenylphosphine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'