Learn More
Bis(4-chlorobutyl) Ether, TRC
Product Code. 30343350
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30343350 1 g 1g
1 options
Product Code. 30343350 Supplier Toronto Research Chemicals Supplier No. TRCB4186301G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(4-chlorobutyl) Ether
CAS 6334-96-9
Molecular Formula C8H16Cl2O
Quantity 1 g
Synonym 1,1'-Oxybis[4-chlorobutane], 1,9-Dichloro-5-oxanonane, 4,4'-Dichlorobutyl Ether, 4,4'-Dichlorodibutyl Ether, NSC 26978
InChI Formula InChI=1S/C8H16Cl2O/c9-5-1-3-7-11-8-4-2-6-10/h1-8H2
SMILES ClCCCCOCCCCCl
IUPAC Name 1-chloro-4-(4-chlorobutoxy)butane
Molecular Weight (g/mol) 199.12
Formula Weight 198.0578