Läs mer
Bis(4-cyanophenyl)methanol, TRC
Produktkod. 30135306
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135306 25 mg
30138537 5 mg
30011951 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135306 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB42285025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134521-16-7
Molekylformel C15 H10 N2 O
Molekylvikt (g/mol) 234.25
Synonym Benzonitrile, 4,4'-(hydroxymethylene)bis-, 4,4'-(Hydroxymethylene)bis[benzonitrile], 4,4'-Methanolbisbenzonitrile, 4-[(4-Cyanophenyl)(hydroxy)methyl]benzonitrile, Bis(4-cyanophenyl)methanol, Bis(p-cyanophenyl)methanol, CGP 44645
IUPAC-namn 4-[(4-cyanophenyl)-hydroxymethyl]benzonitrile
LEDER OC(c1ccc(cc1)C#N)c2ccc(cc2)C#N
Specifikationer
Kemiskt namn eller material Bis(4-cyanophenyl)methanol
Kvantitet 25 mg
InChI formel InChI=1S/C15H10N2O/c16-9-11-1-5-13(6-2-11)15(18)14-7-3-12(10-17)4-8-14/h1-8,15,18H
Formel vikt 234.0793