Läs mer
Bis(4-hydroxyphenyl) Sulfone -13C12, TRC
Produktkod. 30009504
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009504 10 mg
30003758 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB44739310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1991267-29-8
Molekylformel 13C12 H10 O4 S
Molekylvikt (g/mol) 262.18
Synonym 4,4'-Sulfonylbis[phenol-13C6], 1,1'-Sulfonylbis[4-hydroxybenzene-13C6], 4,4'-Bisphenol S 13C12, 4,4'-Dihydroxydiphenyl sulfone 13C12, Bis(4-hydroxyphenyl) sulfone 13C12, Bis(p-hydroxyphenyl) sulfone 13C12, Bisphenol S 13C12, p,p'-Dihydroxydiphenyl sulfone 13C12
IUPAC-namn 4-(4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)sulfonyl(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
LEDER O[13c]1[13cH][13cH][13c]([13cH][13cH]1)S(=O)(=O)[13c]2[13cH][13cH][13c](O)[13cH][13cH]2
Specifikationer
Kemiskt namn eller material Bisphenol S ring-13C12
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H10O4S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8,13-14H/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1
Formel vikt 262.0702
Procent renhet >95%