Läs mer
Bis-(4-nitrophenyl)amine, TRC
Produktkod. 30010714
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010714 50 mg
30006498 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB50611550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1821-27-8
Molekylformel C12 H9 N3 O4
Molekylvikt (g/mol) 259.22
IUPAC-namn 4-nitro-N-(4-nitrophenyl)aniline
LEDER [O-][N+](=O)c1ccc(Nc2ccc(cc2)[N+](=O)[O-])cc1
Specifikationer
Kemiskt namn eller material Bis-(4-nitrophenyl)amine
Kvantitet 50 mg
InChI formel InChI=1S/C12H9N3O4/c16-14(17)11-5-1-9(2-6-11)13-10-3-7-12(8-4-10)15(18)19/h1-8,13H
Formel vikt 259.0593