missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-nitrophenyl) carbonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Bis(4-nitrophenyl) carbonate is used as a reagent for preparation of symmetrical and unsymmetrical urea and 4-nitrophenyl esters of N-protected amino acids. It acts as a peptide coupling reagent. It finds application as a reagent for the preparation of carbamate linked cytosines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(4-nitrophenyl) carbonate is used as a reagent for preparation of symmetrical and unsymmetrical urea and 4-nitrophenyl esters of N-protected amino acids. It acts as a peptide coupling reagent. It finds application as a reagent for the preparation of carbamate linked cytosines.
Solubility
Soluble in chloroform and tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
Bis(4-nitrophenyl) carbonate is used as a reagent for preparation of symmetrical and unsymmetrical urea and 4-nitrophenyl esters of N-protected amino acids. It acts as a peptide coupling reagent. It finds application as a reagent for the preparation of carbamate linked cytosines.
Solubility
Soluble in chloroform and tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
Specifications
Melting Point | 136°C to 139°C |
Quantity | 5 g |
Beilstein | 1892897 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform and tetrahydrofuran. |
Formula Weight | 304.21 |
Percent Purity | 98% |
Chemical Name or Material | Bis(4-nitrophenyl) carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'