Läs mer
Bis(4-nitrophenyl) Phenyl Phosphate, TRC
Produktkod. 30330921
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30330921 1 g
1 options
Produktkod. 30330921 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5964601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(4-nitrophenyl)phenyl Phosphate
CAS 38873-96-0
Molekylformel C18 H13 N2 O8 P
Kvantitet 1 g
Synonym Phosphoric acid, bis(4-nitrophenyl) phenyl ester, Bis(4-nitrophenyl)phenyl Phosphate
InChI formel InChI=1S/C18H13N2O8P/c21-19(22)14-6-10-17(11-7-14)27-29(25,26-16-4-2-1-3-5-16)28-18-12-8-15(9-13-18)20(23)24/h1-13H
LEDER [O-][N+](=O)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccc(cc3)[N+](=O)[O-])cc1
IUPAC-namn bis(4-nitrophenyl) phenyl phosphate
Molekylvikt (g/mol) 416.2782
Formel vikt 416.041