Läs mer
Bis(4-nitrophenyl) sulfone, 95%, Thermo Scientific™
Produktkod. 11489347
Change view
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11489347 25 g 25g
11479347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11489347 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21397.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1156-50-9
Molekylformel C12H8N2O6S
Molekylvikt (g/mol) 308.264
MDL-nummer MFCD00010447
InChI-nyckel BVHNGWRPAFKGFP-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl sulfone, 4-nitrophenyl sulfone, 4,4'-dinitrodiphenyl sulfone, bis 4-nitrophenyl sulphone, bis p-nitrophenyl sulfone, 4,4'-dinitrodiphenylsulfone, 1,1'-sulfonylbis 4-nitrobenzene, sulfone, bis p-nitrophenyl, benzene, 1,1'-sulfonylbis 4-nitro-9ci
PubChem CID 70871
IUPAC-namn 1-nitro-4-(4-nitrophenyl)sulfonylbenzene
LEDER C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Bis(4-nitrophenyl) sulfone
Smältpunkt 252°C to 255°C
Odör Odorless
Kvantitet 25 g
Formel vikt 308.27
Procent renhet 95%