Läs mer
Bis(butoxyethyl)chlorophosphate, TRC
Produktkod. 30339333
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30339333 25 mg
1 options
Produktkod. 30339333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB41081525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(butoxyethyl)chlorophosphate
CAS 20222-73-5
Molekylformel C12H26ClO5P
Kvantitet 25 mg
Synonym Bis(2-butoxyethyl) Phosphorochloridate, Bis(2-butoxyethyl) Ester Phosphorochloridic Acid
InChI formel InChI=1S/C12H26ClO5P/c1-3-5-7-15-9-11-17-19(13,14)18-12-10-16-8-6-4-2/h3-12H2,1-2H3
LEDER CCCCOCCOP(=O)(Cl)OCCOCCCC
IUPAC-namn 1-[2-[2-butoxyethoxy(chloro)phosphoryl]oxyethoxy]butane
Molekylvikt (g/mol) 316.76
Formel vikt 316.1206