missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7081464
Click to view available options
Quantity:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1271-19-8
C10H10Cl2Ti-2
248.957
MFCD00003723
MKNXBRLZBFVUPV-UHFFFAOYSA-L
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
124040768
cyclopenta-1,3-diene;titanium(2+);dichloride
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1271-19-8
C10H10Cl2Ti-2
248.957
MFCD00003723
MKNXBRLZBFVUPV-UHFFFAOYSA-L
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
124040768
cyclopenta-1,3-diene;titanium(2+);dichloride
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, air.
TRUSTED_SUSTAINABILITY
Melting Point ∼285°C (decomposition)
Density 1.6
Odor Odorless
Quantity 10 g
Sensitivity Air and moisture sensitive
Merck Index 14,9482
Solubility Information Slightly soluble in water
Formula Weight 249
Percent Purity 97%
Chemical Name or Material Bis (cyclopentadienyl)titanium dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'