missing translation for 'onlineSavingsMsg'
Läs mer

Bis(diethylamino)chlorophosphine, 94%

Produktkod. 11470097
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11470097 25 g 25g
11460097 5 g 5g
11480097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11470097 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21019.14
Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 685-83-6 | C8H20ClN2P | 210.686 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 685-83-6
Molekylformel C8H20ClN2P
Molekylvikt (g/mol) 210.686
MDL-nummer MFCD00015493
InChI-nyckel JVEHJSIFWIIFHM-UHFFFAOYSA-N
Synonym bis diethylamino chlorophosphine, bis diethylamino phosphorchloridite, n,n,n',n'-tetraethylphosphorodiamidous chloride, bdapcd, chloro diethylamino phosphanyl diethylamine, pubchem6475, phosphorodiamidous chloride, tetraethyl, chlorobis diethylamino phosphine, bisdiethylamino-chloro-phosphine, chloro-bis diethylamino-phosphine
PubChem CID 2733351
IUPAC-namn N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine
LEDER CCN(CC)P(N(CC)CC)Cl

Specifikationer

Kemiskt namn eller material Bis(diethylamino)chlorophosphine
Densitet 1.002
Kokpunkt 55°C to 57°C (0.2 mmHg)
Flampunkt 101°C (214°F)
Odör Acrid
Linjär formel [(C2H5)2N]2PCl
Kvantitet 25 g
UN-nummer UN3265
Känslighet Air and moisture sensitive
Formel vikt 210.69
Procent renhet 94%
Visa mer Visa mindre

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.