Läs mer
Bis(diethylamino)chlorophosphine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Bis(diethylamino)chlorophosphine |
| CAS | 685-83-6 |
| Molekylformel | C8H20ClN2P |
| Kvantitet | 5 g |
| Synonym | N,N,N',N'-Tetraethylphosphorodiamidous Chloride, Bis(N,N-diethylamino)chlorophosphine, Bis(diethylamino)chlorophosphane, Chlorobis(diethylamino)phosphine, Tetraethyldiamidophosphorous Chloride, Tetraethyldiaminochlorophosphite, Tetraethylphosphorodiamidous Chloride |
| InChI formel | InChI=1S/C8H20ClN2P/c1-5-10(6-2)12(9)11(7-3)8-4/h5-8H2,1-4H3 |
| LEDER | CCN(CC)P(Cl)N(CC)CC |
| IUPAC-namn | N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine |
| Molekylvikt (g/mol) | 210.68 |
| Formel vikt | 210.1053 |