Läs mer
(Bis(diphenylmethyl)ether), TRC
Produktkod. 17953915
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17953915 50 mg
17955055 25 mg
17974905 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17953915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB43034550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 574-42-5
Molekylformel C26 H22 O
Molekylvikt (g/mol) 350.45
Synonym [Oxybis(methanetriyl)]tetrabenzene, Bis(diphenylmethyl) Ether, Dimenhydrinate Imp. K (EP)
IUPAC-namn [benzhydryloxy(phenyl)methyl]benzene
LEDER O(C(c1ccccc1)c2ccccc2)C(c3ccccc3)c4ccccc4
Specifikationer
Kemiskt namn eller material Bis(diphenylmethyl) Ether
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H22O/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22)27-26(23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20,25-26H
Formel vikt 350.1671
Procent renhet >95%