missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(ethylcyclopentadienyl)magnesium, 98+%

Brand:  Alfa Aesar™ 038597.06

Additional Details : CAS Number : 114460-02-5 Weight : 0.00500kg Transport : UN number : 3392 Chem class : 4.2 Pack group : I

Quantity 5g

 View more versions of this product

Product Code. 15471487

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »


Specifications

Chemical Identifiers

Bis(ethylcyclopentadienyl)magnesium
114460-02-5
MFCD00151375
UN3392

Specifications

Bis(ethylcyclopentadienyl)magnesium
114460-02-5
Liquid
MFCD00151375
1.91g/mL
Air & Moisture Sensitive
210.6
Ampule
≥98%
UN3392
5g
Videos
SDS