Läs mer
Bis(heptadecafluorooctyl)phosphinic Acid, TRC
Produktkod. 30342166
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30342166 100 mg
1 options
Produktkod. 30342166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB443745100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(heptadecafluorooctyl)phosphinic Acid
CAS 40143-79-1
Molekylformel C16HF34O2P
Kvantitet 100 mg
Synonym Phosphinic acid, P,P-bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)- (ACI), P,P-Bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)phosphinic acid (ACI), P,P-Bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)phosphinic Acid, Phosphinic acid, bis(heptadecafluorooctyl)- (9CI), Bis(perfluorooctyl)phosphinic acid
InChI formel InChI=1S/C16HF34O2P/c17-1(18,5(25,26)9(33,34)13(41,42)43)3(21,22)7(29,30)11(37,38)15(47,48)53(51,52)16(49,50)12(39,40)8(31,32)4(23,24)2(19,20)6(27,28)10(35,36)14(44,45)46/h(H,51,52)
LEDER OP(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn bis(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)phosphinic acid
Molekylvikt (g/mol) 902.1
Formel vikt 901.9171