Läs mer
Bis(m-cresyl) p-Cresyl Phosphate (>90%), TRC
Produktkod. 30334529
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30334529 250 mg
1 options
Produktkod. 30334529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB419225250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(m-cresyl) p-Cresyl Phosphate
CAS 72016-32-1
Molekylformel C21H21O4P
Kvantitet 250 mg
Synonym Phosphoric Acid Bis(3-methylphenyl) 4-Methylphenyl Ester, Di-m-cresyl p-Cresyl Phosphate
InChI formel InChI=1S/C21H21O4P/c1-16-10-12-19(13-11-16)23-26(22,24-20-8-4-6-17(2)14-20)25-21-9-5-7-18(3)15-21/h4-15H,1-3H3
LEDER Cc1ccc(OP(=O)(Oc2cccc(C)c2)Oc3cccc(C)c3)cc1
IUPAC-namn bis(3-methylphenyl) (4-methylphenyl) phosphate
Molekylvikt (g/mol) 368.36
Formel vikt 368.1177