Läs mer
Bis(maltolato)oxovanadium(IV), TRC
Produktkod. 30344393
Change view
Kvantitet:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30344393 10 g
30339418 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30344393 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB47850010G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(maltolato)oxovanadium(IV)
CAS 38213-69-3
Molekylformel C12 H10 O7 V
Kvantitet 10 g
Synonym Vanadium, bis[3-(hydroxy-κO)-2-methyl-4H-pyran-4-onato-κO4]oxo-, (SP-5-31)-, Vanadium, bis(3-hydroxy-2-methyl-4H-pyran-4-onato-O3,O4)oxo-, (SP-5-31)-, (SP-5-31)-Bis[3-(hydroxy-κO)-2-methyl-4H-pyran-4-onato-κO4]oxovanadium, 4H-Pyran-4-one, 3-hydroxy-2-methyl-, vanadium complex, Bis(maltolato)oxovanadium, Bis(maltolato)oxovanadium(IV)
InChI formel InChI=1S/2C6H6O3.O.V/c2*1-4-6(8)5(7)2-3-9-4;;/h2*2-3,8H,1H3;;/q;;;+2/p-2
LEDER CC1=C(O[V](=O)OC2=C(C)OC=CC2=O)C(=O)C=CO1
IUPAC-namn bis[(2-methyl-4-oxopyran-3-yl)oxy]-oxovanadium
Molekylvikt (g/mol) 317.15
Formel vikt 316.9866