Läs mer
Bis-N-Cbz-Lisdexamphetamine, TRC
Produktkod. 30372362
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30372362 100 mg
1 options
Produktkod. 30372362 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD418252100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dibenzyl ((R)-6-oxo-6-(((S)-1-phenylpropan-2-yl)amino)hexane-1,5-diyl)dicarbamate
CAS 5002-57-3
Molekylformel C31 H37 N3 O5
Kvantitet 100 mg
Synonym Dibenzyl ((R)-6-oxo-6-(((S)-1-phenylpropan-2-yl)amino)hexane-1,5-diyl)dicarbamate, Bis-N-Cbz-Lisdexamphetamine, Z-Lys(Z)-Dextroamphetamine
InChI formel InChI=1S/C31H37N3O5/c1-24(21-25-13-5-2-6-14-25)33-29(35)28(34-31(37)39-23-27-17-9-4-10-18-27)19-11-12-20-32-30(36)38-22-26-15-7-3-8-16-26/h2-10,13-18,24,28H,11-12,19-23H2,1H3,(H,32,36)(H,33,35)(H,34,37)/t24-,28+/m0/s1
LEDER C[C@@H](Cc1ccccc1)NC(=O)[C@@H](CCCCNC(=O)OCc2ccccc2)NC(=O)OCc3ccccc3
IUPAC-namn benzyl N-[(2R)-1-oxo-6-(phenylmethoxycarbonylamino)-1-[[(2S)-1-phenylpropan-2-yl]amino]hexan-2-yl]carbamate
Molekylvikt (g/mol) 531.643
Formel vikt 531.273