missing translation for 'onlineSavingsMsg'
Läs mer

Bis(nonafluorobutyl)phosphinic Acid (>90%), TRC

Produktkod. 30134002
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134002 10 mg
30010536 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30134002 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB50427010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52299-25-9
Molekylformel C8HF18O2P
Molekylvikt (g/mol) 502.04
Synonym P-Bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)phosphinic Acid
IUPAC-namn bis(perfluorobutyl)phosphinic acid
LEDER OP(C(F)(C(F)(C(F)(C(F)(F)F)F)F)F)(C(F)(C(F)(C(F)(C(F)(F)F)F)F)F)=O

Specifications

Kemiskt namn eller material Bis(nonafluorobutyl)phosphinic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C8HF18O2P/c9-1(10,5(17,18)19)3(13,14)7(23,24)29(27,28)8(25,26)4(15,16)2(11,12)6(20,21)22/h(H,27,28)
Formel vikt 501.94
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.