Läs mer
Bis(perfluorohexyl)phosphinic Acid Sodium Salt, TRC
Produktkod. 30179720
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30179720 500 mg
1 options
Produktkod. 30179720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB443740500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70609-44-8
Molekylformel C12 F26 O2 P . Na
Molekylvikt (g/mol) 724.05
Synonym 6:6 PFPi, 6:6 PhiA, Phosphinic acid, bis(tridecafluorohexyl)-, sodium salt (9CI)
IUPAC-namn sodium;bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)phosphinate
LEDER [Na+].[O-]P(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material Bis(perfluorohexyl)phosphinic acid sodium salt
Kvantitet 500 mg
InChI formel InChI=1S/C12HF26O2P.Na/c13-1(14,5(21,22)9(29,30)31)3(17,18)7(25,26)11(35,36)41(39,40)12(37,38)8(27,28)4(19,20)2(15,16)6(23,24)10(32,33)34;/h(H,39,40);/q;+1/p-1
Formel vikt 723.9118