Läs mer
Bis(perfluorohexyl)phosphinic Acid, TRC
Produktkod. 30345965
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30345965 100 mg
1 options
Produktkod. 30345965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB443743100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(perfluorohexyl)phosphinic Acid
CAS 40143-77-9
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H F26 O2 P
Kvantitet 100 mg
Synonym Phosphinic acid, P,P-bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)-, Phosphinic acid, bis(tridecafluorohexyl)- (9CI), P,P-Bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)phosphinic acid, Bis(perfluorohexyl)phosphinic acid, Bis(tridecafluorohexyl)phosphinic acid, 6:6 PhiA
InChI formel InChI=1S/C12HF26O2P/c13-1(14,5(21,22)9(29,30)31)3(17,18)7(25,26)11(35,36)41(39,40)12(37,38)8(27,28)4(19,20)2(15,16)6(23,24)10(32,33)34/h(H,39,40)
LEDER OP(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)phosphinic acid
Molekylvikt (g/mol) 702.07
Formel vikt 701.9299