missing translation for 'onlineSavingsMsg'
Learn More

Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-3586404
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
GMLMZLGOMMIIMA-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
This item is not returnable. View return policy
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
GMLMZLGOMMIIMA-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280°C
Color Yellow to Orange
Elemental Analysis 94% min. (ex Carbon)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 786.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Bis[tri(o-tolyl)phosphine]palladium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bis¢tri(o-tolyl)phosphine!palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'