Learn More
Bis(tri-o-tolylphosphine)palladium(0), Pd 14.9%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Bis(tri-o-tolylphosphine)palladium(0) is used as a catalyst for palladium-catalyzed cross-coupling reactions, Kumada coupling and Heck coupling. It is also used in the amination and arylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220°C to 225°C |
Color | White to Yellow |
Assay Percent Range | Pd 14.9% |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 715.16 |
Physical Form | Crystalline |
Chemical Name or Material | Bis(tri-o-tolylphosphine)palladium(0) |