missing translation for 'onlineSavingsMsg'
Learn More

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals

Catalog Number p-2689351
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2712-78-9
C10H5F6IO4
430.03
MFCD00009672
PEZNEXFPRSOYPL-UHFFFAOYSA-N
bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
102317
[phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F
This item is not returnable. View return policy
2712-78-9
C10H5F6IO4
430.03
MFCD00009672
PEZNEXFPRSOYPL-UHFFFAOYSA-N
bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
102317
[phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 125°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula (CF3CO2)2IC6H5
Quantity 10 g
Fieser 09,54
Merck Index 15, 7406
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvent
Formula Weight 430.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material [Bis(trifluoroacetoxy)iodo]benzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • ¢Bis(trifluoroacetoxy)iodo!benzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'