Läs mer
Biscarfentrazone, TRC
Produktkod. 30334974
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334974 5 mg
30334567 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30334974 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1622905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bipyrazone
CAS 1622908-18-2
Molekylformel C20 H19 F3 N4 O5 S
Kvantitet 5 mg
Synonym 1,3-Dimethyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]-1H-pyrazol-5-yl 1,3-dimethyl-1H-pyrazole-4-carboxylate, 1,3-Dimethyl-1H-pyrazole-4-carboxylic acid 1,3-dimethyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]-1H-pyrazol-5-yl ester, Bipyrazone
InChI formel InChI=1S/C20H19F3N4O5S/c1-10-14(9-26(3)24-10)19(29)32-18-16(11(2)25-27(18)4)17(28)13-7-6-12(20(21,22)23)8-15(13)33(5,30)31/h6-9H,1-5H3
LEDER Cc1nn(C)cc1C(=O)Oc2c(C(=O)c3ccc(cc3S(=O)(=O)C)C(F)(F)F)c(C)nn2C
IUPAC-namn [2,5-dimethyl-4-[2-methylsulfonyl-4-(trifluoromethyl)benzoyl]pyrazol-3-yl] 1,3-dimethylpyrazole-4-carboxylate
Molekylvikt (g/mol) 484.45
Formel vikt 484.1028