missing translation for 'onlineSavingsMsg'
Learn More

Bisindolylmaleimide I, MedChemExpress

Product Code. 16435898
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MedChemExpress Bisindolylmaleimide I (GF109203X) is a highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor with a Ki of 14 nM.

Bisindolylmaleimide I (GF109203X) is a highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor with a Ki of 14 nM.

Specifications

Chemical Name or Material Bisindolylmaleimide I
CAS 133052-90-1
Color Red
Purity Grade Notes Research
Molecular Formula C25H24N4O2
Quantity 10 mg
Synonym GF109203X Go 6850
Solubility Information DMSO : ≥ 32 mg/mL (77.58 mM)
SMILES O=C(C(C1=CN(CCCN(C)C)C2=C1C=CC=C2)=C3C4=CNC5=C4C=CC=C5)NC3=O
Molecular Weight (g/mol) 412.48
Formula Weight 412.48
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.