missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth 2-ethylhexanoate acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 638.61 |
Percent Purity | 92% (typically) |
Physical Form | Liquid |
Chemical Name or Material | Bismuth 2-ethylhexanoate, In 2-ethylhexanoic acid |
Safety and Handling
Product Identifier
- Bismuth 2-ethylhexanoate, typically 92%, in 2-ethylhexanoic acid
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'