Läs mer
Bismuth Subgallate, TRC
Produktkod. 30345534
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30345534 1 g
1 options
Produktkod. 30345534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB0163641G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bismuth Subgallate
CAS 99-26-3
Molekylformel C7H5BiO6
Kvantitet 1 g
Synonym 2,7-Dihydroxy-1,3,2-benzodioxabismole-5-carboxylic Acid, Basic Bismuth Gallate, Bismuth Gallate Basic, Bismuth(III) Subgallate, Dermatol, Devrom
InChI formel InChI=1S/C7H6O5.Bi.H2O/c8-4-1-3(7(11)12)2-5(9)6(4)10;;/h1-2,8-10H,(H,11,12);;1H2/q;+3;/p-3
LEDER O[Bi]1Oc2cc(cc(O)c2O1)C(=O)O
IUPAC-namn 2,4-dihydroxy-1,3,2-benzodioxabismole-6-carboxylic acid
Molekylvikt (g/mol) 394.09
Formel vikt 393.989