Läs mer
Bisoctyl Dimethyl Ammonium Chloride, TRC
Produktkod. 30138574
Change view
Kvantitet:
100 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138574 2.5 g
18062420 1 g
18051010 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138574 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5088802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5538-94-3
Molekylformel C18 H40 N . Cl
Molekylvikt (g/mol) 305.97
IUPAC-namn dimethyl(dioctyl)azanium;chloride
LEDER [Cl-].CCCCCCCC[N+](C)(C)CCCCCCCC
Specifikationer
Kemiskt namn eller material Dimethyldioctylammonium chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
Synonym 1-Octanaminium, N,N-dimethyl-N-octyl-, chloride (9CI)
InChI formel InChI=1S/C18H40N.ClH/c1-5-7-9-11-13-15-17-19(3,4)18-16-14-12-10-8-6-2;/h5-18H2,1-4H3;1H/q+1;/p-1
Formel vikt 305.2849
Procent renhet >95%