Läs mer
Bisphenol A Bis(2-hydroxyethyl) Ether, TRC
Produktkod. 30007975
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007975 500 mg
30011966 250 mg
30011810 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB398130500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 901-44-0
Molekylformel C19 H24 O4
Molekylvikt (g/mol) 316.39
Synonym 2,2'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))diethanol, 2,2'-[(1-Methylethylidene)bis(4,1-phenyleneoxy)]bis[ethanol], 1,1'-Isopropylidenebis(p-phenyleneoxy-2-ethanol), 1,4-Di-(β-hydroxyethyl)bisphenol A, 2,2-Bis(4-β-hydroxyethoxyphenyl)propane, 2,2-Bis[4-(2-hydroxyethoxy)phenyl]propane, 2,2-Bis[p-(2-hydroxyethoxy)phenyl]propane, 2,2-Bis[p-(β-hydroxyethoxy)phenyl]propane, 2,2'-[Isopropylidenebis(p-phenyleneoxy)]diethanol, 4,4'-Bis(hydroxyethoxy)diphenyldimethylmethane, 4,4'-Bis(hydroxyethyl)bisphenol A, AE 2, AE 2 (diol), BA 2, BA 2 Glycol, BSA 20, Bisphenol A bis(β-hydroxyethyl) Ether, Bisphenol A Ethylene Oxide Adduct (1:2), Bisphenol A-Ethylene oxide 1:2 mol Adduct, Bisphenol A-Ethylene Oxide Adduct, Dianol 22, Diethoxy Bisphenol A, Diethoxylated Bisphenol A, Ethylene oxide-bisphenol A (2:1) Adduct, Koremul BSA 20, NSC 60933, Newpol BPE 20, Newpol BPE 20T, Simulsol BPLE;
IUPAC-namn 2-[4-[2-[4-(2-hydroxyethoxy)phenyl]propan-2-yl]phenoxy]ethanol
LEDER CC(C)(c1ccc(OCCO)cc1)c2ccc(OCCO)cc2
Specifikationer
Kemiskt namn eller material Bisphenol A Bis(2-hydroxyethyl) Ether
Kvantitet 500 mg
InChI formel InChI=1S/C19H24O4/c1-19(2,15-3-7-17(8-4-15)22-13-11-20)16-5-9-18(10-6-16)23-14-12-21/h3-10,20-21H,11-14H2,1-2H3
Formel vikt 316.1675