Läs mer
Bisphenol F-13C6, TRC
Produktkod. 30041050
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041050 1 mg
30041278 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30041050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5195581MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1410794-06-7
Molekylformel 13C6 C7 H12 O2
Molekylvikt (g/mol) 206.19
Synonym Phenol-1,2,3,4,5,6-13C6, 4-[(4-hydroxyphenyl)methyl]-, 4-[(4-Hydroxyphenyl)methyl]phenol-1,2,3,4,5,6-13C6, 4,4'-Methylenediphenol-1,2,3,4,5,6-13C6, 1-(4-Hydroxyphenyl)-1-(4-hydroxyphenyl-1,2,3,4,5,6-13C6)methane, Bisphenol F (phenyl-13C6)
IUPAC-namn 4-[(4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)methyl]phenol
LEDER O[13c]1[13cH][13cH][13c](Cc2ccc(O)cc2)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material Bisphenol F-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C13H12O2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8,14-15H,9H2/i1+1,2+1,5+1,6+1,10+1,12+1
Formel vikt 206.1039
Procent renhet >95%