Läs mer
Bisphenol G, TRC
Produktkod. 30135591
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135591 1 g
17953385 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135591 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5194501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127-54-8
Molekylformel C21 H28 O2
Molekylvikt (g/mol) 312.45
Synonym Phenol, 4,4'-isopropylidenebis[2-isopropyl- (6CI,7CI,8CI), 4,4'-(1-Methylethylidene)bis[2-(1-methylethyl)phenol], 2,2-Bis(4-hydroxy-3-isopropylphenyl)propane, 4,4'-Isopropylidenebis(2-isopropylphenol), Bis-OIPP-A, Bisphenol G, NSC 21526
IUPAC-namn 4-[2-(4-hydroxy-3-propan-2-ylphenyl)propan-2-yl]-2-propan-2-ylphenol
LEDER CC(C)c1cc(ccc1O)C(C)(C)c2ccc(O)c(c2)C(C)C
Specifikationer
Kemiskt namn eller material Bisphenol G
Kvantitet 1 g
InChI formel InChI=1S/C21H28O2/c1-13(2)17-11-15(7-9-19(17)22)21(5,6)16-8-10-20(23)18(12-16)14(3)4/h7-14,22-23H,1-6H3
Formel vikt 312.2089