Läs mer
Bisphenol Z, TRC
Produktkod. 30132615
Change view
Kvantitet:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132615 50 g
30134345 10 g
30011431 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB51965050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 843-55-0
Molekylformel C18 H20 O2
Molekylvikt (g/mol) 268.35
Synonym Phenol, 4,4'-cyclohexylidenedi- (6CI,8CI), 4,4'-Cyclohexylidenebis[phenol], 1,1-Bis(4-hydroxyphenyl)cyclohexane, 1,1-Bis(p-Hydroxyphenyl)cyclohexane, 4,4'-Cyclohexylidenediphenol, Antigene W, Bis-Z, Bisphenol Z, Dian C, NSC 29881, NSC 50761
IUPAC-namn 4-[1-(4-hydroxyphenyl)cyclohexyl]phenol
LEDER Oc1ccc(cc1)C2(CCCCC2)c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material Bisphenol Z
Kvantitet 50 g
InChI formel InChI=1S/C18H20O2/c19-16-8-4-14(5-9-16)18(12-2-1-3-13-18)15-6-10-17(20)11-7-15/h4-11,19-20H,1-3,12-13H2
Formel vikt 268.1463