Läs mer
Bivalirudin Trifluoroacetic Acid Salt, TRC
Produktkod. 30007957
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007957 10 mg
30003453 100 mg
30007578 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007957 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB59163010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1124915-67-8
Molekylformel C98H138N24O33 . (C2HF3O2)x
Molekylvikt (g/mol) 2180.2911402
Synonym D-Phenylalanyl-L-prolyl-L-arginyl-L-prolylglycylglycylglycylglycyl-L-asparaginylglycyl-L-α-aspartyl-L-phenylalanyl-L-α-glutamyl-L-α-glutamyl-L-isoleucyl-L-prolyl-L-α-glutamyl-L-α-glutamyl-L-tyrosyl-L-leucine Trifluoroacetic Acid Salt, Angiomax, Angiox, BG 8967, Hirulog, Hirulog1, Kangio;
LEDER [H][H]C([C@@H](NN1CCC[C@H]1C(N[C@@H](CCCNC(N)=N)C(N2CCC[C@H]2C(NCC(NCC(NCC(NCC(N[C@@H](CC(N)=O)C(N[C]C(N[C@H](C(N[C@H](C(N[C@@H](CCC(O)=O)C(N[C@@H](CCC(O)=O)C(N[C@H](C(N3CCC[C@H]3C(N[C@@H](CCC(O)=O)C(N[C@@H](CCC(O)=O)C(N[C@H](C(N[C@@H](CC(C)C)C(O)=O)=O)CC4=CC=C(O)C=C4)=O)=O)=O)=O)[C@@H](C)CC)=O)=O)=O)CC5=CC=CC=C5)=O)CC(O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)CC6=CC=CC=C6)=O.OC(C(F)(F)F)=O
Specifikationer
Kemiskt namn eller material Bivalirudin Trifluoroacetic Acid Salt
Kvantitet 10 mg
Formel vikt 2292.9787