missing translation for 'onlineSavingsMsg'
Läs mer

BIX 02189, TRC

Produktkod. 30334645
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334645 5 mg
30331624 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Product Code. 30334645 Supplier Toronto Research Chemicals Supplier No. TRCB5917655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (3Z)-3-[((3-[(Dimethylamino)methyl]phenyl)amino)phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide
CAS 1265916-41-3
Molekylformel C27H28N4O2
Kvantitet 5 mg
Synonym BIX 02189, (3Z)-3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide
InChI formel InChI=1S/C27H28N4O2/c1-30(2)17-18-9-8-12-21(15-18)28-25(19-10-6-5-7-11-19)24-22-14-13-20(27(33)31(3)4)16-23(22)29-26(24)32/h5-16,29,32H,17H2,1-4H3
LEDER CN(C)Cc1cccc(N\C(=C\2/C(=O)Nc3cc(ccc23)C(=O)N(C)C)\c4ccccc4)c1
IUPAC-namn 3-[N-[3-[(dimethylamino)methyl]phenyl]-C-phenylcarbonimidoyl]-2-hydroxy-N,N-dimethyl-1H-indole-6-carboxamide
Molekylvikt (g/mol) 440.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.