Läs mer
BIX02189, MedChemExpress
Produktkod. 16447697
Encompass_Preferred
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16447697 5 mg
16441207 50 mg
16425736 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16447697 Leverantör Medchem Express Leverantörsnummer HY12056
MedChemExpress BIX02189 is a potent and selective MEK5 inhibitor with an IC50 of 1.5 nM. BIX02189 also inhibits ERK5 catalytic activity with an IC50 of 59 nM.
BIX02189 is a potent and selective MEK5 inhibitor with an IC50 of 1.5 nM. BIX02189 also inhibits ERK5 catalytic activity with an IC50 of 59 nM.
Specifikationer
Kemiskt namn eller material BIX02189
Fysisk form Solid
CAS 1265916-41-3
Färg Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C27H28N4O2
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : ≥ 49.4 mg/mL (112.14 mM)
LEDER O=C(N(C)C)C1=CC(NC2=O)=C(/C2=C(C3=CC=CC=C3)/NC4=CC=CC(CN(C)C)=C4)C=C1.[(Z)]
Molekylvikt (g/mol) 440.54
Formel vikt 440.54
Procent renhet 99.57%