Learn More
BLACE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559786
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HAWLYLTRHF PWSPFPHGGW TDTSEPCVLE TLGGSSLAAL WGNSLWVQSS GACAFCVYES LIEQSLPNER FEELLLGPSP GEVM Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 23%.
BLACE Polyclonal antibody specifically detects BLACE in Human samples. It is validated for Immunohistochemistry.Specifications
BLACE | |
Polyclonal | |
Unconjugated | |
BLACE | |
B-cell acute lymphoblastic leukemia expressed; BLACE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338436 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A4D250 | |
BLACE | |
Recombinant protein corresponding to Human BLACE | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only