Learn More
Uvex Black/Orange PU 65021 Low Shoe - Width 10
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine the limits for safety shoes; range in protection classes S3 and S1 P
1770.00 SEK
Specifications
Color | Black, Orange |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17280691
|
Uvex
6502135 |
35 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17290691
|
Uvex
6502136 |
36 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17200701
|
Uvex
6502137 |
37 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17210701
|
Uvex
6502138 |
38 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17220701
|
Uvex
6502139 |
39 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17230701
|
Uvex
6502140 |
40 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17240701
|
Uvex
6502141 |
41 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17250701
|
Uvex
6502142 |
42 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17260701
|
Uvex
6502143 |
43 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17270701
|
Uvex
6502144 |
44 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17280701
|
Uvex
6502145 |
45 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17290701
|
Uvex
6502146 |
46 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17200711
|
Uvex
6502147 |
47 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17210711
|
Uvex
6502148 |
48 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17220711
|
Uvex
6502149 |
49 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17230711
|
Uvex
6502150 |
50 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17240711
|
Uvex
6502151 |
51 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17250711
|
Uvex
6502152 |
52 |
1770.00 SEK
1 pair |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Shoe 6502.1 S3 SRC
- Sporty, multifunctional safety shoes
- Lightweight and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side, no thermal conductivity and anti-magnetic properties
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, which does not restrict flexibility
- Made with new developed technology for exceptional wearer comfort
- Uvex Climazone technology for optimum wearer comfort
- Breathable, water-resistant, leather upper material
- Distance mesh lining provides optimum breathability
- Comes with elastic laces for quick individual adjustment
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Uvex topshield scuffcap to protect the upper and extend its life
- Fulfills ESD requirements: resistance to ground Kleiner of 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6502.1
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S3 |