Learn More
Linden™ Polyethylene Bucket
For general purpose carrying and storage applications. Linden™ Polyethylene Buckets come with a strong handle with hand grip.
Brand: Linden™ H-TOOL-BUCKET-14L
Additional Details : Weight : 1.00000kg
Description
- With pouring spout
- Durable construction
- 14L capacity
- Black in color
Specifications
Bucket | |
Yes | |
Black |
14 L | |
Polyethylene | |
1 Ea. |