Läs mer
Blasticidin S Hydrochloride, TRC
Produktkod. 30330062
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30330062 25 mg
1 options
Produktkod. 30330062 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB59280025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Blasticidin S Hydrochloride
CAS 3513-03-9
Molekylformel C17 H26 N8 O5 . Cl H
Kvantitet 25 mg
Synonym Blasticidin S, monohydrochloride (8CI), β-D-erythro-Hex-2-enopyranuronic acid, 4-[[(3S)-3-amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-, monohydrochloride (9CI), β-D-erythro-Hex-2-enopyranuronic acid, 4-[[3-amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-, monohydrochloride, (S)-, Blasticidin S hydrochloride, β-D-erythro-Hex-2-enopyranuronic acid, 4-[[(3S)-3-amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-, hydrochloride (1:1)
InChI formel InChI=1S/C17H26N8O5.ClH/c1-24(16(20)21)6-4-9(18)8-12(26)22-10-2-3-13(30-14(10)15(27)28)25-7-5-11(19)23-17(25)29;/h2-3,5,7,9-10,13-14H,4,6,8,18H2,1H3,(H3,20,21)(H,22,26)(H,27,28)(H2,19,23,29);1H/t9-,10-,13+,14-;/m0./s1
LEDER Cl.CN(CC[C@H](N)CC(=O)N[C@H]1C=C[C@@H](O[C@@H]1C(=O)O)N2C=CC(=NC2=O)N)C(=N)N
IUPAC-namn (2S,3S,6R)-3-[[(3S)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 458.9
Formel vikt 458.1793