Learn More
BRAND™ BLAUBRAND™ Micropipettes
BLAUBRAND™ Micropipettes with DE-M and CE markings.
Brand: BRAND™ 708727
Additional Details : Weight : 1.00000kg
Description
- DE-M marking
- Calibrated to contain (TC, In)
- Color code to ISO for clear identification
- CE-marked according to IVD-Directive 98/79 CE
Specifications
± 0.25% | |
250/Pk | |
2 x red |
DE-M | |
Jar | |
40 μL |