Learn More
Thermo Scientific™ Blocker™ BSA Blocking Buffers
Block membranes, plates and other samples with these ready-to-use 10X PBS or TBS solutions of bovine serum albumin for Western blot, ELISA and other detection methods.
1334.00 SEK - 1430.00 SEK
Specifications
Applications ELISA, IHC, WB
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10341944 Thermo Scientific™
37525
Blocker BSA in PBS (10X) 1430.00 SEK
200mL View alternative products
Save Up to 
»
EU Stock 25
Add to basket
11889310 Thermo Scientific™
37520
Blocker BSA in TBS (10X) 1334.00 SEK
125mL View alternative products
Save Up to 
»
EU Stock 16
Add to basket
Description
Description
These blocking buffers are 10% (w/v) solutions of high-quality BSA that are useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. Blocker BSA Buffers are most frequently diluted 10-fold (to 1% BSA) for initial testing, but other buffer concentrations can be beneficial for specific systems. Blocker BSA is usually more effective than nonfat milk blocking buffers for biotin-avidin systems because it contains a single purified protein that is devoid endogenous biotin.
Highlights:
  • Purified protein – 10% solutions of high-quality bovine serum albumin; a single purified protein provides fewer chances of cross-reaction with assay components than serum or milk solutions
  • Convenient – concentrated formulation saves storage space and can be diluted easily to obtain optimal blocking results for specific applications
  • Easy to use – no waiting for powder to dissolve with this ready-to-dilute liquid concentrate
  • Flexible – available in PBS and TBS formulations to suit a variety of applications
Specifications
Specifications
ELISA, IHC, WB