Läs mer
Blonanserin, TRC
Produktkod. 30334140
Change view
Kvantitet:
5 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334140 100 mg
30329884 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30334140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB595850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Blonanserin
CAS 132810-10-7
Anteckningar om renhetsgrad HPLC
Molekylformel C23 H30 F N3
Kvantitet 100 mg
Synonym 2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydro-cycloocta[b]pyridine, Lonasen, AD 5423
InChI formel InChI=1S/C23H30FN3/c1-2-26-13-15-27(16-14-26)23-17-21(18-9-11-19(24)12-10-18)20-7-5-3-4-6-8-22(20)25-23/h9-12,17H,2-8,13-16H2,1H3
LEDER CCN1CCN(CC1)c2cc(c3CCCCCCc3n2)c4ccc(F)cc4
IUPAC-namn 2-(4-ethylpiperazin-1-yl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
Molekylvikt (g/mol) 367.5
Formel vikt 367.2424