Läs mer
BD Vacutainer™ Blood Transfer Device
Produktkod. 12937736
Change view
Förpackningsstorlek:
Paket med 200
Produktkod. Förpackningsstorlek
12937736 Paket med 200
1 options
Produktkod. 12937736 Leverantör BD Leverantörsnummer 36481000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Enables needleless transfer of blood. BD Biosciences™ Vacutainer™ Blood Transfer Device With luer adapter.
  • For blood transfer
  • Sterilized with EtO
TRUSTED_SUSTAINABILITY
Specifikationer
Inkluderar With luer adapter
Produkttyp Blood Transfer Device
Sterilitet Sterile