missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BLOTTO and Tween 20 in PBS, with 0.02% sodium azide

Blocking reagent for immunohistochemistry experiments

Brand:  Alfa Aesar™ J62304.AK

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15464709

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Specifications

Specifications

Blocking reagent for immunohistochemistry experiments
Blocking Buffer
−30° to −10°C
Liquid
250mL
Videos
SDS