Learn More
Tenak™ Blue Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand:  Tenak™ TE22055
Additional Details : Weight : 3.20000kg
 View more versions of this product
Product Code. 15230524
  • 1300.00 SEK / Pack of 36
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Specifications
Blue
Square
133 x 133 x 75 mm
Cardboard
Cryo Box