Learn More
Uvex Blue PU 85328 Boot - Width 11
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 and S2
1315.00 SEK - 1355.00 SEK
Specifications
Color | Blue |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17224971
|
Uvex
8532835 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17234971
|
Uvex
8532836 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17244971
|
Uvex
8532837 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17254971
|
Uvex
8532838 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17264971
|
Uvex
8532839 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17274971
|
Uvex
8532840 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17284971
|
Uvex
8532841 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17294971
|
Uvex
8532842 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17204981
|
Uvex
8532843 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17214981
|
Uvex
8532844 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17224981
|
Uvex
8532845 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17234981
|
Uvex
8532846 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17244981
|
Uvex
8532847 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17254981
|
Uvex
8532848 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17264981
|
Uvex
8532849 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17274981
|
Uvex
8532850 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17284981
|
Uvex
8532851 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17294981
|
Uvex
8532852 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 1 Boot 8532.8 S1 SRC
- Sporty, multifunctional safety shoe
- Especially lightweight and flexible
- 100% metal-free Uvex Xenova™ toecap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Made with new developed last technology for extended wearing comfort
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, perforated, high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Soft padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8532.8
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Standard width | |
Yes | |
S1 |