Läs mer
SI Analytics™ BlueLine pH Combination Electrode with Conical Membrane
Handla allt SI Analytics Produkter
Kontakttyper:
BNC
DIN, 4 mm banankontakt, 4 mm Banana Plug
DÅN
Kontakthuvud
Metrohm-kontakt
Modell:
Blueline 11 PH
Blueline 12 PH
Blueline 14 PH
Blueline 14 PH-id
Blueline 17 PH
Blueline 17 pH-R
Blueline 18 PH
Förpackningsstorlek:
Varje
Beskrivning
- Glass bodied with robust cone shaped membrane, Ag/AgCl reference system
- For universal applications
- Electrolyte KCl 3 mol/l
- With fixed cable 1 m, Ø = 5 mm, DIN or BNC connector and 4 mm banana connector
- SCHOTT™ Instruments BlueLine 14 pH (DIN-plug + banana, fixed cable)
Specifikationer
Specifikationer
| Kontakttyper | DIN, 4 mm banankontakt, 4 mm Banana Plug |
| Korsning | Platinum Diaphragm |
| Material | Glass |
| pH-intervall | 0 to 14 |
| Typ | Combination pH Electrode |
| Intern referens | Ag/AgCl |
| Membran | Cone |
| Modell | Blueline 14 PH |
| Temperaturområde (metriskt). | -5°C to +100°C |
| Längd (metrisk) | 120 mm |
| Visa mer |