Läs mer
Invitrogen™ BMI-1 Monoclonal Antibody (282CT3.7.6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA537525
Produktkod. 17107118
  • 5120.00 SEK / 400µL
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
BMI-1 Monoclonal Antibody for Western Blot, ICC/IF
Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
BMI-1
Monoclonal
Unconjugated
BMI1
AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1
Mouse
Protein G
RUO
648
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
282CT3.7.6
PBS with 0.09% sodium azide; pH 7.4
P35226
BMI1
BMI1 recombinant protein.
400 μL
Primary
Human
Antibody
IgG1 κ