missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22858-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
BMP10 is a member of the TGF-beta family of growth factors. Data suggest that the similar protein in mouse plays an important role in trabeculation of the embryonic heart. In human, this protein may signal through receptor serine/threonine kinases.Specifications
BMP10 | |
Polyclonal | |
Unconjugated | |
BMP10 | |
BMP 10, BMP10, bone morphogenetic protein 10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27302 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95393 | |
BMP10 | |
BMP10 Fusion Protein Ag18746 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |